Assay Details
| Gene Symbol: Acad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172678.3 | NP_766266.3 | 8 - 9 | 1002 | 66 | |
| GenBank mRNA | AK028571.1 | - | 7 - 8 | 1716 | 66 | |
| AK031820.1 | - | 8 - 9 | 934 | 66 | ||
| AK075984.1 | - | 8 - 9 | 921 | 66 | ||
| AK145423.1 | - | 8 - 9 | 910 | 66 | ||
| AK203069.1 | - | 2 - 3 | 67 | |||
| AK206153.1 | - | 2 - 3 | 216 | 66 | ||
| AK217282.1 | - | 2 - 3 | 226 | 66 | ||
| BC004737.1 | - | 2 - 3 | 248 | 66 | ||
| BC031137.1 | - | 8 - 9 | 938 | 4971848 | 66 | |
| BC032213.1 | - | 8 - 9 | 943 | 5323723 | 66 | |
| BC033277.1 | - | 8 - 9 | 923 | 4241041 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acad9
Genomic Map