Assay Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009316.1 | NP_033342.1 | 4 - 5 | 501 | 94 | |
| NM_172688.3 | NP_766276.1 | 4 - 5 | 501 | 94 | ||
| GenBank mRNA | AK046565.1 | - | 4 - 5 | 467 | 94 | |
| AK053911.1 | - | 4 - 5 | 487 | 94 | ||
| AK137046.1 | - | 4 - 5 | 502 | 94 | ||
| AK145302.1 | - | 4 - 5 | 512 | 94 | ||
| AK159277.1 | - | 4 - 5 | 465 | 94 | ||
| AK211827.1 | - | 2 - 3 | 99 | 94 | ||
| BC006665.1 | - | 4 - 5 | 492 | 3499247 | 94 | |
| D76446.1 | - | 4 - 5 | 501 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k7
Genomic Map