Assay Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009316.1 | NP_033342.1 | 6 - 7 | 66 | ||
| NM_172688.3 | NP_766276.1 | 6 - 7 | 66 | |||
| GenBank mRNA | AK053911.1 | - | 6 - 7 | 66 | ||
| AK137046.1 | - | 6 - 7 | 66 | |||
| AK159277.1 | - | 6 - 7 | 66 | |||
| BC006665.1 | - | 6 - 7 | 756 | 3499247 | 66 | |
| D76446.1 | - | 6 - 7 | 765 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k7
Genomic Map