Assay Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009316.1 | NP_033342.1 | 15 - 16 | 1674 | 74 | |
| NM_172688.3 | NP_766276.1 | 14 - 15 | 1593 | 74 | ||
| GenBank mRNA | AK046565.1 | - | 14 - 15 | 1559 | 74 | |
| AK053911.1 | - | 14 - 15 | 1579 | 74 | ||
| AK133642.1 | - | 8 - 9 | 720 | 74 | ||
| AK137046.1 | - | 14 - 15 | 1594 | 74 | ||
| AK159277.1 | - | 14 - 15 | 1557 | 74 | ||
| BC006665.1 | - | 15 - 16 | 1665 | 3499247 | 74 | |
| D76446.1 | - | 14 - 15 | 1593 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k7
Genomic Map