Assay Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 11 - 12 | 1771 | 75 | |
| GenBank mRNA | AK049305.1 | - | 2 - 3 | 362 | 75 | |
| AK052595.1 | - | 7 - 8 | 1025 | 75 | ||
| AK128929.1 | - | 11 - 12 | 1706 | 75 | ||
| AK155930.1 | - | 11 - 12 | 1707 | 75 | ||
| AY553221.1 | - | 11 - 12 | 1647 | 75 | ||
| BC027369.1 | - | 11 - 12 | 1750 | 75 | ||
| BC106095.1 | - | 11 - 12 | 1691 | 75 | ||
| BC127163.1 | - | 11 - 12 | 1637 | 75 | ||
| BC152540.1 | - | 11 - 12 | 1639 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx58
Genomic Map