Assay Details
| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172693.3 | NP_766281.1 | 4 - 5 | 921 | 118 | |
| GenBank mRNA | AB175680.1 | - | 4 - 5 | 901 | 118 | |
| AK033638.1 | - | 4 - 5 | 922 | 118 | ||
| AK042133.1 | - | 4 - 5 | 954 | 118 | ||
| AK043977.1 | - | 3 - 4 | 781 | 118 | ||
| AK082014.1 | - | 4 - 5 | 967 | 118 | ||
| AK156751.1 | - | 4 - 5 | 951 | 118 | ||
| BC024475.1 | - | 3 - 4 | 753 | 118 | ||
| BC025639.1 | - | 3 - 4 | 643 | 118 | ||
| BC056425.1 | - | 4 - 5 | 932 | 5716909 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt12
Genomic Map