Assay Details
| Gene Symbol: Serinc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253386.1 | NP_001240315.1 | 7 - 8 | 960 | 57 | |
| NM_172702.3 | NP_766290.2 | 7 - 8 | 955 | 57 | ||
| GenBank mRNA | AK075863.1 | - | 7 - 8 | 923 | 57 | |
| AK158914.1 | - | 7 - 8 | 961 | 57 | ||
| BC031720.1 | - | 7 - 8 | 936 | 4209575 | 57 | |
| CB055666.1 | - | 2 - 3 | 149 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serinc2
Genomic Map