Assay Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172710.3 | NP_766298.2 | 10 - 11 | 1943 | 58 | |
| GenBank mRNA | AK030487.1 | - | 10 - 11 | 1808 | 58 | |
| AK037220.1 | - | 10 - 11 | 1807 | 58 | ||
| AK122362.1 | - | 10 - 11 | 1943 | 58 | ||
| BC094607.1 | - | 9 - 10 | 1488 | 6837048 | 58 | |
| BC117915.1 | - | 9 - 10 | 1444 | 40097358 | 58 | |
| BC117916.1 | - | 9 - 10 | 1446 | 40097359 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sel1l3
Genomic Map