Assay Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172710.3 | NP_766298.2 | 11 - 12 | 2119 | 98 | |
| GenBank mRNA | AK030487.1 | - | 11 - 12 | 1984 | 98 | |
| AK037220.1 | - | 11 - 12 | 1983 | 98 | ||
| AK122362.1 | - | 11 - 12 | 2119 | 98 | ||
| BC094607.1 | - | 10 - 11 | 1664 | 6837048 | 98 | |
| BC117915.1 | - | 10 - 11 | 1620 | 40097358 | 98 | |
| BC117916.1 | - | 10 - 11 | 1622 | 40097359 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sel1l3
Genomic Map