Assay Details
| Gene Symbol: Lin54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115010.1 | NP_001108482.1 | 3 - 4 | 1115 | 74 | |
| NM_172714.4 | NP_766302.2 | 3 - 4 | 983 | 74 | ||
| GenBank mRNA | AK029288.1 | - | 3 - 4 | 330 | 74 | |
| AK030894.1 | - | 3 - 4 | 1117 | 74 | ||
| AK134320.1 | - | 3 - 4 | 981 | 74 | ||
| AK220225.1 | - | 3 - 4 | 978 | 74 | ||
| BC106126.1 | - | 2 - 3 | 619 | 74 | ||
| BC125580.1 | - | 1 - 2 | 110 | 74 | ||
| BC145025.1 | - | 1 - 2 | 110 | 9053573 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC145025.1 transcripts.
Set Membership: |
|

Lin54
Genomic Map