Assay Details
| Gene Symbol: Lin54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115010.1 | NP_001108482.1 | 7 - 8 | 1750 | 108 | |
| NM_172714.4 | NP_766302.2 | 7 - 8 | 1618 | 108 | ||
| GenBank mRNA | AK029288.1 | - | 8 - 9 | 1064 | 108 | |
| AK030894.1 | - | 7 - 8 | 1752 | 108 | ||
| AK134320.1 | - | 7 - 8 | 1616 | 108 | ||
| AK220225.1 | - | 7 - 8 | 1613 | 108 | ||
| BC106126.1 | - | 6 - 7 | 1254 | 108 | ||
| BC125580.1 | - | 5 - 6 | 745 | 108 | ||
| BC145025.1 | - | 5 - 6 | 745 | 9053573 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lin54
Genomic Map