Assay Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172716.4 | NP_766304.2 | 2 - 3 | 155 | 65 | |
| GenBank mRNA | AK033609.1 | - | 2 - 3 | 230 | 65 | |
| AK085574.1 | - | 2 - 3 | 211 | 65 | ||
| AK089091.1 | - | 2 - 3 | 156 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK033609.1,AK089091.1,NM_172716.4 transcripts.
Set Membership: |
|

Pcgf3
Genomic Map