Assay Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172716.4 | NP_766304.2 | 3 - 4 | 298 | 102 | |
| GenBank mRNA | AK033609.1 | - | 3 - 4 | 373 | 102 | |
| AK085574.1 | - | 3 - 4 | 354 | 102 | ||
| AK089091.1 | - | 3 - 4 | 299 | 102 | ||
| BC049266.1 | - | 165 | 5371040 | 102 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK033609.1,AK089091.1,BC049266.1,NM_172716.4 transcripts.
Set Membership: |
|

Pcgf3
Genomic Map