Assay Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172716.4 | NP_766304.2 | 4 - 5 | 421 | 76 | |
| GenBank mRNA | AK033609.1 | - | 4 - 5 | 496 | 76 | |
| AK085574.1 | - | 4 - 5 | 477 | 76 | ||
| AK089091.1 | - | 4 - 5 | 422 | 76 | ||
More Information
Set Membership: |
|

Pcgf3
Genomic Map