Assay Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172718.3 | NP_766306.2 | 23 - 24 | 3115 | 66 | |
| NR_027934.1 | 14 - 15 | 2302 | 66 | |||
| GenBank mRNA | AK053555.1 | - | 23 - 24 | 3049 | 66 | |
| AK167632.1 | - | 14 - 15 | 2266 | 66 | ||
| BC138049.1 | - | 23 - 24 | 3041 | 8861068 | 66 | |
| BC138050.1 | - | 23 - 24 | 3041 | 8861069 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm1
Genomic Map