Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 41 - 42 | 5377 | 105 | |
| GenBank mRNA | AB093217.1 | - | 17 - 18 | 2570 | 105 | |
| AK077013.1 | - | 5 - 6 | 699 | 105 | ||
| AK090137.1 | - | 41 - 42 | 5374 | 105 | ||
| AK147257.1 | - | 41 - 42 | 5370 | 105 | ||
| BC056933.1 | - | 7 - 8 | 1009 | 105 | ||
| BC150735.1 | - | 41 - 42 | 5380 | 9087646 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcn1l1
Genomic Map