Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 45 - 46 | 6025 | 69 | |
| GenBank mRNA | AB093217.1 | - | 21 - 22 | 3218 | 69 | |
| AK077013.1 | - | 9 - 10 | 1347 | 69 | ||
| AK090137.1 | - | 45 - 46 | 6022 | 69 | ||
| AK147257.1 | - | 45 - 46 | 6018 | 69 | ||
| BC056933.1 | - | 11 - 12 | 1657 | 69 | ||
| BC068244.1 | - | 1 - 2 | 279 | 69 | ||
| BC150735.1 | - | 45 - 46 | 6028 | 9087646 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC068244.1 transcripts.
Set Membership: |
|

Gcn1l1
Genomic Map