Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 8 - 9 | 803 | 85 | |
| GenBank mRNA | AK049325.1 | - | 8 - 9 | 804 | 85 | |
| AK052060.1 | - | 8 - 9 | 791 | 85 | ||
| AK149758.1 | - | 8 - 9 | 804 | 85 | ||
| AK165175.1 | - | 8 - 9 | 794 | 85 | ||
| AK167405.1 | - | 8 - 9 | 821 | 85 | ||
| AK167491.1 | - | 8 - 9 | 821 | 85 | ||
| BC028868.1 | - | 6 - 7 | 583 | 85 | ||
| BC037595.1 | - | 8 - 9 | 784 | 85 | ||
| BC055480.1 | - | 7 - 8 | 705 | 85 | ||
| BC082314.1 | - | 8 - 9 | 789 | 85 | ||
| BC106182.1 | - | 8 - 9 | 778 | 85 | ||
| BC132172.1 | - | 8 - 9 | 801 | 40130449 | 85 | |
| BC138093.1 | - | 8 - 9 | 801 | 8861112 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map