Assay Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172748.2 | NP_766336.2 | 9 - 10 | 2017 | 82 | |
| GenBank mRNA | AK028405.1 | - | 9 - 10 | 1784 | 82 | |
| AK031431.1 | - | 7 - 8 | 1410 | 82 | ||
| AK162161.1 | - | 3 - 4 | 396 | 82 | ||
| BC053109.1 | - | 5 - 6 | 1010 | 82 | ||
| BC059812.1 | - | 9 - 10 | 2017 | 6401846 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl19
Genomic Map