Assay Details
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078520.1 | - | 2 - 3 | 4049 | 74 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | NM_001081268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | NM_172749.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | AK078520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | AK129112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | BC068300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | NM_001081268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | NM_172749.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | AK078520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | AK129112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466773 | BC068300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022789 | NM_172749.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022789 | AK078520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022789 | AK129112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022789 | BC068300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022789 | BC079907.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92986 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8547 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145084 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022789 | NM_172749.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022789 | AK078520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022789 | AK129112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022789 | BC068300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022789 | BC079907.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | NM_001081268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | NM_172749.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | AK078520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | AK129112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | BC068300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | NM_001081268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | NM_172749.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | AK078520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | AK129112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466773 | BC068300.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507842 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01632043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145084 | - | - |
| Whole Mouse Genome G4122A | A_51_P466773 | NM_001081268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | NM_172749.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | AK078520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | AK129112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | BC068300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | NM_001081268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | NM_172749.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | AK078520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | AK129112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466773 | BC068300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40641 | AK028847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40641 | NM_172749.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40641 | BC079907.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121020 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06623673 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06623674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05951059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06422255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06623673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507845 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557811 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557815 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459901_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439212_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557808 | - | - |
| Mouse Genome 430 2.0 Array | 1459901_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459901_at | - | - |
| Mouse Genome 430 2.0 Array | 1439212_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439212_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233905_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233905_at | - | - |
| Mouse Expression Set 430 | 1439212_at | - | - |
| Mouse Expression Set 430 | 1439212_PM_at | - | - |
| Mouse Expression Set 430 | 1459901_PM_at | - | - |
| Mouse Expression Set 430 | 1459901_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439212_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459901_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459901_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439212_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 433821 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5360184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5360184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360184 | - | - |
| mouse-6 v1.1 (Illumina) | scl0233905.3_263-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0233905.3_263-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2991339 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706460 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2991339 | - | - |

Zfp646
Genomic Map