Assay Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172760.3 | NP_766348.3 | 4 - 5 | 396 | 91 | |
| GenBank mRNA | AK037067.1 | - | 4 - 5 | 389 | 91 | |
| BC058752.1 | - | 4 - 5 | 379 | 6485393 | 91 | |
| BC138887.1 | - | 4 - 5 | 353 | 8861909 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295933 | BC058752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295933 | BC138887.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295933 | BC018516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295933 | BC026617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2295933 | AK157875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC058752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC138887.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC018516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC026617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | NM_172760.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC058752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC138887.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC018516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | BC026617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P114620 | NM_172760.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516892 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162829 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC058752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC138887.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC018516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC026617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | NM_172760.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC058752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC138887.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC018516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | BC026617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P114620 | NM_172760.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295933 | BC058752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295933 | BC138887.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295933 | BC018516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295933 | BC026617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2295933 | AK157875.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132905 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162829 | - | - |
| Whole Mouse Genome G4122A | A_52_P114620 | BC058752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC138887.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC018516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC026617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | NM_172760.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC058752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC138887.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC018516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | BC026617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114620 | NM_172760.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475948 | AK048129.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475948 | BC027048.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475948 | BC023859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475948 | BC026649.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P475948 | BC027030.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120589 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130446 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162829 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082811 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07082813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07082836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07082817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07082819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07082853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07082811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574711 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574710 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574707 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434489_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574694 | - | - |
| Mouse Genome 430 2.0 Array | 1434489_at | - | - |
| Mouse Genome 430 2.0 Array | 1434489_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234683_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234683_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800009546.mm.1 | NM_172760.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009546.mm.1 | NM_172760.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009546.mm.1 | TC0800001213.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434489_PM_at | - | - |
| Mouse Expression Set 430 | 1434489_at | - | - |
| Murine Genome U74v2 Set | 167433_at | - | - |
| Murine Genome U74v2 Set | 167433_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434489_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434489_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434489_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434489_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 436539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2640019 | - | - |
| MouseWG-6 v2 Expression BeadChip | 110372 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110372 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2640019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693636 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219244 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219244 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2693636 | - | - |
| mouse-6 v1.1 (Illumina) | scl33417.13.1_5-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0234683.19_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640019 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110372 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0234683.19_25-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33417.13.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640019 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110372 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2693636 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219244 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2693636 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219244 | - | - |

Elmo3
Genomic Map