Assay Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172787.2 | NP_766375.1 | 18 - 19 | 1981 | 114 | |
| GenBank mRNA | AK030412.1 | - | 5 - 6 | 579 | 114 | |
| AK045667.1 | - | 18 - 19 | 1982 | 114 | ||
| AK129443.1 | - | 14 - 15 | 1526 | 114 | ||
| BC082309.1 | - | 17 - 18 | 1851 | 30356672 | 114 | |
| BC085192.1 | - | 17 - 18 | 2086 | 30694487 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl3
Genomic Map