Assay Details
| Gene Symbol: Enox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253759.1 | NP_001240688.1 | 9 - 10 | 1340 | 74 | |
| NM_172813.3 | NP_766401.1 | 9 - 10 | 1340 | 74 | ||
| GenBank mRNA | AK045500.1 | - | 9 - 10 | 1258 | 74 | |
| AK051821.1 | - | 9 - 10 | 1288 | 74 | ||
| AK082388.1 | - | 9 - 10 | 1341 | 74 | ||
| AK162001.1 | - | 9 - 10 | 1289 | 74 | ||
| BC052062.1 | - | 6 - 7 | 851 | 5703279 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Enox1
Genomic Map