Assay Details
| Gene Symbol: Lmln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172823.2 | NP_766411.1 | 6 - 7 | 722 | 93 | |
| GenBank mRNA | AK030463.1 | - | 6 - 7 | 723 | 93 | |
| BC111899.1 | - | 6 - 7 | 723 | 40062192 | 93 | |
| BC112428.1 | - | 6 - 7 | 723 | 40062196 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmln
Genomic Map