Assay Details
| Gene Symbol: Lmln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172823.2 | NP_766411.1 | 10 - 11 | 1147 | 61 | |
| GenBank mRNA | AK030463.1 | - | 10 - 11 | 1148 | 61 | |
| BC111899.1 | - | 10 - 11 | 1148 | 40062192 | 61 | |
| BC112428.1 | - | 10 - 11 | 1148 | 40062196 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmln
Genomic Map