Assay Details
| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159649.1 | NP_001153121.1 | 4 - 5 | 693 | 90 | |
| NM_172842.3 | NP_766430.3 | 4 - 5 | 595 | 90 | ||
| GenBank mRNA | AK040834.1 | - | 4 - 5 | 503 | 90 | |
| AK088503.1 | - | 4 - 5 | 446 | 90 | ||
| AY090785.1 | - | 3 - 4 | 321 | 90 | ||
| BC042765.1 | - | 4 - 5 | 595 | 4188712 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lax1
Genomic Map