Assay Details
| Gene Symbol: Fmo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172844.2 | NP_766432.1 | 1 - 2 | 91 | 113 | |
| GenBank mRNA | AK029219.1 | - | 1 - 2 | 92 | 113 | |
| AK161224.1 | - | 1 - 2 | 91 | 113 | ||
| BC112405.1 | - | 1 - 2 | 81 | 40060428 | 113 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK029219.1,AK161224.1,BC112405.1,NM_172844.2 transcripts.
Set Membership: |
|

Fmo9
Genomic Map