Assay Details
| Gene Symbol: Fmo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172844.2 | NP_766432.1 | 7 - 8 | 1041 | 94 | |
| GenBank mRNA | AK029219.1 | - | 7 - 8 | 1042 | 94 | |
| AK161224.1 | - | 7 - 8 | 1041 | 94 | ||
| BC111885.1 | - | 5 - 6 | 770 | 40060432 | 94 | |
| BC112405.1 | - | 7 - 8 | 1031 | 40060428 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006837 | NM_172844.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006837 | AK029219.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767866 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119337 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175845 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173337 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006837 | NM_172844.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006837 | AK029219.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20800994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20801019 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00242576 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175845 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173337 | - | - |
| Whole Mouse Genome G4122A | A_51_P415931 | BC112405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415931 | BC111885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415931 | NM_172844.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P415931 | AK029219.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123544 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127058 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173337 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00168023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00168025 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00168030 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00168031 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00528053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00168033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20801067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20800970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00969013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00848418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00528066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00386629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00168033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00168035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00723999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00764077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00104282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00723991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00969009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00969012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00528063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00764075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00969003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20801046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20800965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10359847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10359841 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10359837 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100028461.mm.1 | NM_172844.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028461.mm.1 | NM_172844.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028461.mm.1 | TC0100003473.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 752436 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 60072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 60072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000060072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224512 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224512 | - | - |
| mouse-6 v1.1 (Illumina) | scl15989.10.1_126-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 60072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl15989.10.1_126-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60072 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2864559 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2864559 | - | - |

Fmo9
Genomic Map