Assay Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172852.3 | NP_766440.2 | 1 - 2 | 44 | 66 | |
| GenBank mRNA | AK030670.1 | - | 1 - 2 | 46 | 66 | |
| BC117990.1 | - | 1 - 2 | 34 | 40104505 | 66 | |
| BC117991.1 | - | 1 - 2 | 34 | 66 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC117990.1,BC117991.1,NM_172852.3 transcripts.
Set Membership: |
|

Serpinb13
Genomic Map