Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 12 - 13 | 1820 | 86 | |
| GenBank mRNA | AK030132.1 | - | 12 - 13 | 1822 | 86 | |
| BC092241.1 | - | 10 - 11 | 1074 | 30670567 | 86 | |
| BC144750.1 | - | 11 - 12 | 1203 | 9053298 | 86 | |
| BC150662.1 | - | 12 - 13 | 1297 | 9087573 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570235 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570233 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570234 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570232 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040600 | AK030132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040600 | BC092241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040600 | NM_172857.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | AB063578.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | AK008516.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | AK053445.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | AK143527.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | AK154332.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | BC010447.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | BC034874.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | BC052050.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | BC071271.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | BC096486.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | NM_027248.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932314 | NM_198249.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753804 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331009 | XM_911308.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53451 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174968 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140523 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040600 | AK030132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040600 | BC092241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040600 | NM_172857.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21570233 | - | - |
| Whole Mouse Genome G4122A | A_51_P131281 | AK030132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131281 | BC092241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P131281 | NM_172857.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132391 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02012462 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02012471 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02126677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02126675 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570235 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570232 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570233 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21570234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21570235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02266684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01560927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01506481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01249316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02034117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01684343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02155203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21570222 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486248 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486250 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445191_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486241 | - | - |
| Mouse Genome 430 2.0 Array | 1445191_at | - | - |
| Mouse Genome 430 2.0 Array | 1445191_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 241624_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 241624_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200037059.mm.1 | TC0200004437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200037061.mm.1 | NM_172857.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037061.mm.1 | NM_172857.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037061.mm.1 | TC0200004437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200037060.mm.1 | NM_172857.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037060.mm.1 | NM_172857.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037060.mm.1 | TC0200004437.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1445191_PM_at | - | - |
| Mouse Expression Set 430 | 1445191_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445191_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445191_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 499164 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160260 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160260 | - | - |
| mouse-6 v1.1 (Illumina) | GI_27370297-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160260 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160260 | - | - |

Exd1
Genomic Map