Assay Details
| Gene Symbol: Dzank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172859.2 | NP_766447.2 | 9 - 10 | 1046 | 103 | |
| GenBank mRNA | AK032182.1 | - | 9 - 10 | 1046 | 103 | |
| AK032632.1 | - | 9 - 10 | 1037 | 103 | ||
| AK163994.1 | - | 9 - 10 | 1037 | 103 | ||
| BC141156.1 | - | 9 - 10 | 945 | 9055768 | 103 | |
| BC145464.1 | - | 9 - 10 | 945 | 9054026 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dzank1
Genomic Map