Assay Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172862.3 | NP_766450.2 | 22 - 23 | 9099 | 73 | |
| GenBank mRNA | AB236662.1 | - | 22 - 23 | 9099 | 73 | |
| AJ833643.1 | - | 22 - 23 | 8828 | 73 | ||
| AK031494.1 | - | 12 - 13 | 2001 | 73 | ||
| AK082256.1 | - | 5 - 6 | 1129 | 73 | ||
| BC046388.1 | - | 2 - 3 | 278 | 73 | ||
| BC068185.1 | - | 12 - 13 | 2001 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frem2
Genomic Map