Assay Details
| Gene Symbol: Rgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172866.3 | NP_766454.1 | 3 - 4 | 541 | 74 | |
| GenBank mRNA | AK089288.1 | - | 3 - 4 | 324 | 74 | |
| AK129103.1 | - | 3 - 4 | 327 | 74 | ||
| AK170117.1 | - | 3 - 4 | 338 | 74 | ||
| BC086614.1 | - | 3 - 4 | 347 | 30533256 | 74 | |
| CJ065220.1 | - | 3 - 4 | 545 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgp1
Genomic Map