Assay Details
| Gene Symbol: Podn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285956.1 | NP_001272885.1 | 3 - 4 | 473 | 87 | |
| NM_172874.3 | NP_766462.2 | 3 - 4 | 395 | 87 | ||
| GenBank mRNA | AK034987.1 | - | 3 - 4 | 260 | 87 | |
| AK161018.1 | - | 3 - 4 | 260 | 87 | ||
| AY313606.1 | - | 3 - 4 | 395 | 87 | ||
| BC094340.1 | - | 3 - 4 | 473 | 30609151 | 87 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY313606.1,BC094340.1,NM_001285956.1,NM_172874.3 transcripts.
Set Membership: |
|

Podn
Genomic Map