Assay Details
| Gene Symbol: Tmprss11e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172880.2 | NP_766468.1 | 8 - 9 | 1017 | 92 | |
| GenBank mRNA | AK037235.1 | - | 8 - 9 | 1019 | 92 | |
| AK162701.1 | - | 8 - 9 | 1015 | 92 | ||
| AY787860.1 | - | 8 - 9 | 1025 | 92 | ||
| BC115432.1 | - | 8 - 9 | 1011 | 40094164 | 92 | |
| BC115433.1 | - | 8 - 9 | 1011 | 40094170 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11e
Genomic Map