Assay Details
| Gene Symbol: Mfsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172883.2 | NP_766471.1 | 7 - 8 | 1028 | 71 | |
| GenBank mRNA | AK029021.1 | - | 7 - 8 | 1028 | 71 | |
| AK087226.1 | - | 7 - 8 | 1238 | 71 | ||
| AK171069.1 | - | 6 - 7 | 3660 | 71 | ||
| BC094512.1 | - | 5 - 6 | 640 | 2616131 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd7a
Genomic Map