Assay Details
| Gene Symbol: Tmem132d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172885.2 | NP_766473.1 | 7 - 8 | 2624 | 117 | |
| GenBank mRNA | AB100355.1 | - | 7 - 8 | 2078 | 117 | |
| AK083210.1 | - | 7 - 8 | 2625 | 117 | ||
| BC140953.1 | - | 7 - 8 | 2164 | 9055565 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem132d
Genomic Map