Assay Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172893.3 | NP_766481.2 | 9 - 10 | 1714 | 103 | |
| GenBank mRNA | AK036886.1 | - | 9 - 10 | 1717 | 103 | |
| AK053205.1 | - | 9 - 10 | 1615 | 103 | ||
| AK081528.1 | - | 9 - 10 | 1616 | 103 | ||
| AK156623.1 | - | 9 - 10 | 1583 | 103 | ||
| AK162200.1 | - | 9 - 10 | 1286 | 104 | ||
| AK200035.1 | - | 3 - 4 | 275 | 103 | ||
| AK218645.1 | - | 3 - 4 | 274 | 103 | ||
| BC024579.1 | - | 4 - 5 | 363 | 103 | ||
| BC028906.1 | - | 1 - 2 | 73 | 103 | ||
| BC048927.1 | - | 5 - 6 | 565 | 103 | ||
| BC120733.1 | - | 9 - 10 | 1547 | 40129656 | 103 | |
| BC137645.1 | - | 9 - 10 | 1547 | 8860664 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp12
Genomic Map