Assay Details
| Gene Symbol: Dmkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166173.1 | NP_001159645.1 | 3 - 4 | 782 | 63 | |
| NM_001166174.1 | NP_001159646.1 | 3 - 4 | 782 | 63 | ||
| NM_172899.4 | NP_766487.3 | 3 - 4 | 782 | 63 | ||
| GenBank mRNA | AK081753.1 | - | 3 - 4 | 784 | 63 | |
| AY444557.1 | - | 3 - 4 | 739 | 63 | ||
| AY622963.1 | - | 3 - 4 | 614 | 63 | ||
| BC064724.1 | - | 3 - 4 | 775 | 30435974 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmkn
Genomic Map