Assay Details
| Gene Symbol: Dmkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166173.1 | NP_001159645.1 | 10 - 11 | 1308 | 90 | |
| NM_001166174.1 | NP_001159646.1 | 8 - 9 | 1215 | 90 | ||
| NM_172899.4 | NP_766487.3 | 9 - 10 | 1260 | 90 | ||
| GenBank mRNA | AK081753.1 | - | 10 - 11 | 1310 | 90 | |
| AY444557.1 | - | 8 - 9 | 1172 | 90 | ||
| AY622963.1 | - | 8 - 9 | 1119 | 90 | ||
| BC064724.1 | - | 9 - 10 | 1253 | 30435974 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmkn
Genomic Map