Assay Details
| Gene Symbol: Ovch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172908.3 | NP_766496.2 | 3 - 4 | 286 | 78 | |
| GenBank mRNA | AK033776.1 | - | 2 - 3 | 175 | 78 | |
| BC125284.1 | - | 3 - 4 | 315 | 40129799 | 78 | |
| BC125288.1 | - | 3 - 4 | 315 | 40129803 | 78 | |
| HQ009505.1 | - | 1 - 2 | 39 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ovch2
Genomic Map