Assay Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172909.4 | NP_766497.3 | 1 - 2 | 364 | 78 | |
| GenBank mRNA | AF539744.1 | - | 1 - 2 | 120 | 78 | |
| AK046675.1 | - | 1 - 2 | 365 | 78 | ||
| AK088010.1 | - | 1 - 2 | 351 | 78 | ||
| BC145639.1 | - | 1 - 2 | 90 | 78 | ||
| BC145640.1 | - | 1 - 2 | 90 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989563 | AK088010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989563 | AW494409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989563 | AK046675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989563 | NM_172909.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989563 | AF539744.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640139 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61103 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134114 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137225 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989563 | AK088010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989563 | AW494409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989563 | AK046675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989563 | NM_172909.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989563 | AF539744.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582903 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134114 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137225 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06393905 | - | - |
| Whole Mouse Genome G4122A | A_51_P171382 | AK088010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P171382 | AW494409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P171382 | AK046675.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P171382 | NM_172909.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P171382 | AF539744.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101507 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134114 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06363999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06111727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06195870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06204856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06363995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25582906 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558624 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440808_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455527_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558614 | - | - |
| Mouse Genome 430 2.0 Array | 1440808_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455527_at | - | - |
| Mouse Genome 430 2.0 Array | 1440808_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455527_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244233_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244233_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700017622.mm.1 | TC0700002007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017622.mm.1 | NM_172909.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017622.mm.1 | NM_172909.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017623.mm.1 | TC0700002007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017623.mm.1 | NM_172909.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017623.mm.1 | NM_172909.4 | Refseq |
| Mouse Expression Set 430 | 1440808_x_at | - | - |
| Mouse Expression Set 430 | 1455527_at | - | - |
| Mouse Expression Set 430 | 1440808_PM_x_at | - | - |
| Mouse Expression Set 430 | 1455527_PM_at | - | - |
| Murine Genome U74v2 Set | 133242_at | - | - |
| Murine Genome U74v2 Set | 133242_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455527_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455527_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440808_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440808_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 457910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1770551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227374 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227374 | - | - |
| mouse-6 v1.1 (Illumina) | scl31916.12.1_47-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31916.12.1_47-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2629663 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2629663 | - | - |

Cd163l1
Genomic Map