Assay Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172909.4 | NP_766497.3 | 12 - 13 | 3101 | 60 | |
| GenBank mRNA | AF539744.1 | - | 12 - 13 | 2857 | 60 | |
| AK046675.1 | - | 11 - 12 | 3278 | 60 | ||
| AK088010.1 | - | 11 - 12 | 3094 | 60 | ||
| AW494409.1 | - | 2 - 3 | 60 | |||
| BC145639.1 | - | 10 - 11 | 2926 | 60 | ||
| BC145640.1 | - | 11 - 12 | 2950 | 60 | ||
| BF469445.1 | - | 3 - 4 | 331 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd163l1
Genomic Map