Assay Details
| Gene Symbol: Tox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172913.3 | NP_766501.2 | 3 - 4 | 710 | 69 | |
| GenBank mRNA | AK082602.1 | - | 3 - 4 | 708 | 69 | |
| BC052044.1 | - | 3 - 4 | 436 | 5708447 | 69 | |
| EF410108.1 | - | 574 | 69 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tox3
Genomic Map