Assay Details
| Gene Symbol: Zfp846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172919.2 | NP_766507.2 | 5 - 6 | 724 | 102 | |
| GenBank mRNA | AK033687.1 | - | 6 - 7 | 630 | 102 | |
| BC139282.1 | - | 3 - 4 | 272 | 8862304 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26343013 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26343012 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26343014 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P589664 | AK033687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P589664 | NM_172919.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P589664 | AK033687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P589664 | NM_172919.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | BC139282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | AK033687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | NM_172919.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | AK008693.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | BC139282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | AK033687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | NM_172919.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P52272 | AK008693.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33464 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167164 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139793 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117727 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | BC139282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | AK033687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | NM_172919.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | AK008693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | BC139282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | AK033687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | NM_172919.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P52272 | AK008693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P589664 | AK033687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P589664 | NM_172919.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P589664 | AK033687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P589664 | NM_172919.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26342996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26342995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26342998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26343026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117727 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01880722 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139793 | - | - |
| Whole Mouse Genome G4122A | A_52_P52272 | BC139282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | AK033687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | NM_172919.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | AK008693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | BC139282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | AK033687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | NM_172919.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52272 | AK008693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P589664 | AK033687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P589664 | NM_172919.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P589664 | AK033687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P589664 | NM_172919.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135072 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129412 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117727 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08096194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08096193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08096195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26343013 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26343012 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26343014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26342998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26343030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07642844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08096193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07634523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07757870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08096194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08044172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07734288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07997464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07634521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07656769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08195099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26343025 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583451 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431985_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456023_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583450 | - | - |
| Mouse Genome 430 2.0 Array | 1456023_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431985_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456023_at | - | - |
| Mouse Genome 430 2.0 Array | 1431985_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244721_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244721_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900001056.mm.1 | NM_172919.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001056.mm.1 | NM_172919.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001056.mm.1 | TC0900000173.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900001057.mm.1 | NM_172919.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001057.mm.1 | NM_172919.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001057.mm.1 | TC0900000173.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900001055.mm.1 | NM_172919.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001055.mm.1 | NM_172919.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001055.mm.1 | TC0900000173.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431985_PM_at | - | - |
| Mouse Expression Set 430 | 1456023_at | - | - |
| Mouse Expression Set 430 | 1431985_at | - | - |
| Mouse Expression Set 430 | 1456023_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431985_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431985_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456023_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456023_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 816179 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106200520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1230484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106200520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001230484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255544 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255544 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223827 | - | - |
| mouse-6 v1.1 (Illumina) | scl37245.7_152-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106200520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1230484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37245.7_152-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230484 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106200520 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2776139 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223827 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776139 | - | - |

Zfp846
Genomic Map