Assay Details
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175256.5 | NP_780465.4 | 1 - 2 | 389 | 114 | |
| GenBank mRNA | AK028518.1 | - | 1 - 2 | 390 | 114 | |
| AK163128.1 | - | 1 - 2 | 390 | 114 | ||
| BB639887.1 | - | 1 - 2 | 390 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Heg1
Genomic Map