Assay Details
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028518.1 | - | 2 - 3 | 685 | 97 | |
| AK163128.1 | - | 2 - 3 | 685 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028518.1 | - | 2 - 3 | 685 | 97 | |
| AK163128.1 | - | 2 - 3 | 685 | 97 | ||
Set Membership: |
|