Assay Details
| Gene Symbol: Adck5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172960.3 | NP_766548.2 | 3 - 4 | 390 | 76 | |
| NR_073123.1 | 3 - 4 | 390 | 76 | |||
| GenBank mRNA | AK039217.1 | - | 3 - 4 | 391 | 76 | |
| AK163557.1 | - | 3 - 4 | 338 | 76 | ||
| BC030881.1 | - | 3 - 4 | 370 | 76 | ||
| BC060528.1 | - | 3 - 4 | 279 | 6414642 | 76 | |
| BQ886383.1 | - | 3 - 4 | 313 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC060528.1 transcripts.
Set Membership: |
|

Adck5
Genomic Map