Assay Details
| Gene Symbol: Adck5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172960.3 | NP_766548.2 | 9 - 10 | 1141 | 75 | |
| NR_073123.1 | 9 - 10 | 1136 | 75 | |||
| GenBank mRNA | AK039217.1 | - | 9 - 10 | 1137 | 75 | |
| AK163557.1 | - | 8 - 9 | 1168 | 75 | ||
| BC030881.1 | - | 7 - 8 | 1301 | 75 | ||
| BC060528.1 | - | 9 - 10 | 1025 | 6414642 | 75 | |
More Information
Set Membership: |
|

Adck5
Genomic Map