Assay Details
| Gene Symbol: Inpp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290797.1 | NP_001277726.1 | 16 - 17 | 2025 | 62 | |
| NM_001290798.1 | NP_001277727.1 | 6 - 7 | 904 | 62 | ||
| NM_001290799.1 | NP_001277728.1 | 6 - 7 | 901 | 62 | ||
| NM_030266.4 | NP_084542.2 | 16 - 17 | 2028 | 62 | ||
| NM_172971.3 | NP_766559.2 | 6 - 7 | 904 | 62 | ||
| GenBank mRNA | AK035867.1 | - | 6 - 7 | 905 | 62 | |
| AK149414.1 | - | 15 - 16 | 1935 | 62 | ||
| BC120541.1 | - | 6 - 7 | 900 | 40129464 | 62 | |
| BC144747.1 | - | 6 - 7 | 901 | 62 | ||
| BC144748.1 | - | 6 - 7 | 897 | 9053296 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp4a
Genomic Map